KEY465192865 Key Organics / BIONET

4-chloro-N'-{[(1Z)-3-oxo-1,3-dihydro-2-benzofuran-1-ylidene]methyl}benzohydrazide

  • MDL number MFCD00139642
  • Cas Number : 320424-84-8
  • Molecular Weight : 314.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63204

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-060_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart