KEY465192868 Key Organics / BIONET

3-(2-chlorophenyl)-5-[(E)-2-(phenylamino)ethenyl]-1,2-oxazole-4-carbonitrile

  • MDL number MFCD00127242
  • Cas Number : 320424-89-3
  • Molecular Weight : 321.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63209

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart