KEY465192870 Key Organics / BIONET

3-(2-chlorophenyl)-5-[(E)-2-[(4-methoxyphenyl)amino]ethenyl]-1,2-oxazole-4-carbonitrile

  • MDL number MFCD00127243
  • Cas Number : 320424-91-7
  • Molecular Weight : 351.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1C-070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart