KEY465192871 Key Organics / BIONET

4-{[(E)-2-[3-(2-chlorophenyl)-4-cyano-1,2-oxazol-5-yl]ethenyl]amino}benzoic acid

  • MDL number MFCD00127244
  • Cas Number : 320424-92-8
  • Molecular Weight : 365.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63212

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-072_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart