KEY465192882 Key Organics / BIONET

(4Z)-4-{[(5-chloro-2-hydroxyphenyl)amino]methylidene}-1-methyl-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00138395
  • Cas Number : 320425-04-5
  • Molecular Weight : 319.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63231

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart