KEY465192883 Key Organics / BIONET

(4Z)-1-methyl-4-{[(4-nitrophenyl)amino]methylidene}-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00138399
  • Cas Number : 320425-08-9
  • Molecular Weight : 314.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63235

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-102_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart