KEY465192884 Key Organics / BIONET

methyl 4-({[(4Z)-1-methyl-5-oxo-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}amino)benzoate

  • MDL number MFCD00138401
  • Cas Number : 320425-12-5
  • Molecular Weight : 327.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63239

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-108_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart