KEY465192893 Key Organics / BIONET

methyl 2-({[(4Z)-1-methyl-5-oxo-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-4-ylidene]methyl}amino)acetate

  • MDL number MFCD00138409
  • Cas Number : 320415-85-8
  • Molecular Weight : 265.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-140_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart