KEY465192919 Key Organics / BIONET

7-(3,4-dichlorophenyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00138881
  • Cas Number : 320417-13-8
  • Molecular Weight : 289.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63353

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1D-130_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart