KEY465192923 Key Organics / BIONET

7-[4-(1H-imidazol-1-yl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00138887
  • Cas Number : 320417-27-4
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63361

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1D-138_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart