KEY465192937 Key Organics / BIONET

4-chloro-6-(4-chlorophenyl)-2-(pyridin-3-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139450
  • Cas Number : 320417-58-1
  • Molecular Weight : 327.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1E-035S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart