KEY465192941 Key Organics / BIONET

4-(morpholin-4-yl)-6-phenyl-2-(pyridin-3-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139455
  • Cas Number : 320417-69-4
  • Molecular Weight : 343.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63406

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1E-048S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart