KEY465192946 Key Organics / BIONET

4-(4-chlorophenyl)-6-[(propan-2-yl)amino]-2-(pyridin-3-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139459
  • Cas Number : 320417-88-7
  • Molecular Weight : 349.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63430

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1E-072S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart