KEY465192958 Key Organics / BIONET

4-(4-chlorophenyl)-6-[(prop-2-en-1-yl)amino]-2-(pyridin-2-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139474
  • Cas Number : 320418-17-5
  • Molecular Weight : 347.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63467

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1E-113S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart