KEY465192960 Key Organics / BIONET

4-(4-chlorophenyl)-6-(4-methylpiperazin-1-yl)-2-(pyridin-2-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139476
  • Cas Number : 320418-23-3
  • Molecular Weight : 390.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63473

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1E-122S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart