KEY465192962 Key Organics / BIONET

4-(4-chlorophenyl)-6-(diethylamino)-2-(pyridin-4-yl)pyrimidine-5-carbonitrile

  • MDL number MFCD00139478
  • Cas Number : 320418-26-6
  • Molecular Weight : 363.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63478

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1E-130S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart