KEY465192979 Key Organics / BIONET

2-(pyridin-2-yl)-4-(pyridin-4-yl)pyrimidine-5-carboxylic acid

  • MDL number MFCD01936101
  • Cas Number : 400077-52-3
  • Molecular Weight : 278.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63523

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1E-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart