KEY465192981 Key Organics / BIONET

N-(2,4-dichlorophenyl)-1-{N'-[2-(1H-1,2,4-triazol-1-yl)ethanimidoyl]hydrazinecarbonyl}formamide

  • MDL number MFCD00172022
  • Cas Number : 320418-96-0
  • Molecular Weight : 356.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63534

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1F-002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart