KEY465192982 Key Organics / BIONET

N-methyl-4-[2-(trifluoromethyl)benzoyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD00141847
  • Cas Number : 320418-99-3
  • Molecular Weight : 296.249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63537

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1F-006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart