KEY465192985 Key Organics / BIONET

2,2,2-trichloro-1-[4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrol-2-yl]ethan-1-one

  • MDL number MFCD00141849
  • Cas Number : 320419-19-0
  • Molecular Weight : 399.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63552

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1F-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart