KEY465192992 Key Organics / BIONET

4-hydroxy-6-oxo-1-phenyl-1,6-dihydropyridazine-3-carboxamide

  • MDL number MFCD00141144
  • Cas Number : 121583-00-4
  • Molecular Weight : 231.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1F-057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart