KEY465193004 Key Organics / BIONET

2-(5-cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)ethyl 4-methylbenzoate

  • MDL number MFCD00172939
  • Cas Number : 320420-11-9
  • Molecular Weight : 299.286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63661

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1F-912_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart