KEY465193024 Key Organics / BIONET

3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-4-phenyl-2,5-dihydrofuran-2-one

  • MDL number MFCD00172762
  • Cas Number : 320420-57-3
  • Molecular Weight : 339.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63732

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart