KEY465193037 Key Organics / BIONET

methyl 4-{4-[3-(trifluoromethyl)phenyl]-1,3-thiazol-2-yl}benzoate

  • MDL number MFCD00171763
  • Cas Number : 321430-12-0
  • Molecular Weight : 363.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63858

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-428S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart