KEY465193040 Key Organics / BIONET

2-(prop-2-yn-1-yl)-9-thia-2,4-diazatricyclo[8.4.0.0^{3,8}]tetradeca-1(10),3(8),4,6,11,13-hexaene

  • MDL number MFCD01316361
  • Cas Number : 477864-55-4
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63869

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-445S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart