KEY465193044 Key Organics / BIONET

1-(4-methoxyphenyl)-2-phenyl-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD00232289
  • Cas Number : 51495-00-2
  • Molecular Weight : 317.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63900

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1G-930_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart