KEY465193052 Key Organics / BIONET

1,3-dimethyl-5-({4-[(1Z)-3-oxobut-1-en-1-yl]phenyl}methylidene)-1,3-diazinane-2,4,6-trione

  • MDL number MFCD00141785
  • Cas Number : 321430-64-2
  • Molecular Weight : 312.325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart