KEY465193053 Key Organics / BIONET

(2E)-2-{[(4-chlorophenyl)amino]methylidene}-7-fluoro-3-phenyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD00232244
  • Cas Number : 321430-66-4
  • Molecular Weight : 363.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63931

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart