KEY465193056 Key Organics / BIONET

5-[1-amino-2-(4-chlorophenoxy)ethylidene]-1,3-dimethyl-1,3-diazinane-2,4,6-trione

  • MDL number MFCD00231721
  • Cas Number : 321430-70-0
  • Molecular Weight : 323.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63935

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-019_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart