KEY465193060 Key Organics / BIONET

5-[1-amino-2-(3-chlorophenoxy)ethylidene]-1,3-dimethyl-1,3-diazinane-2,4,6-trione

  • MDL number MFCD00215014
  • Cas Number : 321430-78-8
  • Molecular Weight : 323.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63946

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart