KEY465193061 Key Organics / BIONET

ethyl 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrrole-2-carboxylate

  • MDL number MFCD00231917
  • Cas Number : 321430-87-9
  • Molecular Weight : 318.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63961

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart