KEY465193063 Key Organics / BIONET

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]sulfanyl}butan-2-one

  • MDL number MFCD00231724
  • Cas Number : 321430-99-3
  • Molecular Weight : 283.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63974

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart