KEY465193070 Key Organics / BIONET

5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00172392
  • Cas Number : 103058-77-1
  • Molecular Weight : 271.199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63999

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-326S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart