KEY465193072 Key Organics / BIONET

4-{[(4-fluorophenyl)sulfanyl]methyl}aniline

  • MDL number MFCD00172489
  • Cas Number : 321431-79-2
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-415S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart