KEY465193073 Key Organics / BIONET

3,5-diethyl 6-{[(E)-[(dimethylamino)methylidene]amino]amino}-2-oxo-1,2-dihydropyridine-3,5-dicarboxylate

  • MDL number MFCD00202763
  • Cas Number : 321432-01-3
  • Molecular Weight : 324.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64059

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart