KEY465193082 Key Organics / BIONET

5-[(1E)-1-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazin-1-ylidene}ethyl]-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00172945
  • Cas Number : 321432-20-6
  • Molecular Weight : 375.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64088

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1H-936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart