KEY465193087 Key Organics / BIONET

ethyl 5-({[3-(2-chlorophenyl)-5-methyl-1,2-oxazol-4-yl]formamido}methyl)-4,5-dihydro-1,2-oxazole-3-carboxylate

  • MDL number MFCD00665161
  • Cas Number : 321432-32-0
  • Molecular Weight : 391.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64107

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1H-961_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart