KEY465193090 Key Organics / BIONET

N2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]pyridine-2,6-diamine

  • MDL number MFCD01814583
  • Cas Number : 477864-63-4
  • Molecular Weight : 288.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64125

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1J-007_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart