KEY465193093 Key Organics / BIONET

1-[4-(benzenesulfonyl)phenyl]ethan-1-one

  • MDL number MFCD00173085
  • Cas Number : 65085-83-8
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64134

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1J-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart