KEY465193105 Key Organics / BIONET

1,1-bis(4-chlorophenyl)-2-{[(4-methylphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD00215288
  • Cas Number : 321432-77-3
  • Molecular Weight : 386.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1J-316S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart