KEY465193107 Key Organics / BIONET

1,1-bis(4-chlorophenyl)-2-{[(2-fluorophenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD00215292
  • Cas Number : 321432-80-8
  • Molecular Weight : 390.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1J-321S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart