KEY465193129 Key Organics / BIONET

N-[(E)-2-(benzenesulfonyl)-2-nitroethenyl]-3-chloro-2-methylaniline

  • MDL number MFCD00214845
  • Cas Number : 321433-85-6
  • Molecular Weight : 352.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1K-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart