KEY465193140 Key Organics / BIONET

3-(trifluoromethyl)phenyl 4-methyl-1,2,3-thiadiazole-5-carboxylate

  • MDL number MFCD01314890
  • Cas Number : 477864-72-5
  • Molecular Weight : 288.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64365

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1K-339S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart