KEY465193143 Key Organics / BIONET

N-(5-acetyl-1-benzyl-6-methyl-2-oxo-1,2-dihydropyridin-3-yl)-4-methylbenzamide

  • MDL number MFCD00215438
  • Cas Number : 477864-89-4
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64404

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1K-908_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart