KEY465193144 Key Organics / BIONET

N-{5-acetyl-1-[(furan-2-yl)methyl]-6-methyl-2-oxo-1,2-dihydropyridin-3-yl}-4-chlorobenzamide

  • MDL number MFCD00215441
  • Cas Number : 477864-93-0
  • Molecular Weight : 384.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1K-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart