KEY465193145 Key Organics / BIONET

3-chloro-N-{1-[(furan-2-yl)methyl]-2,5-dioxo-1,2,5,6,7,8-hexahydroquinolin-3-yl}benzamide

  • MDL number MFCD00215442
  • Cas Number : 477864-94-1
  • Molecular Weight : 396.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1K-914_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart