KEY465193152 Key Organics / BIONET

7-[1-(4-tert-butylphenoxy)ethyl]-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD00243604
  • Cas Number : 477865-05-7
  • Molecular Weight : 311.389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64421

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart