KEY465193167 Key Organics / BIONET

methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD00244305
  • Cas Number : 77867-47-1
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64451

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1L-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart