KEY465193178 Key Organics / BIONET

3-{4-[(3-oxobutan-2-yl)oxy]phenoxy}butan-2-one

  • MDL number MFCD00243646
  • Cas Number : 477865-72-8
  • Molecular Weight : 250.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-090_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart