KEY465193185 Key Organics / BIONET

ethyl 6-{[(4-chlorophenyl)methyl]amino}-5-cyano-2-phenylpyridine-3-carboxylate

  • MDL number MFCD00793665
  • Cas Number : 477866-12-9
  • Molecular Weight : 391.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-326S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart